(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%
(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%
(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%
Thermo Scientific Chemicals

(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%

CAS: 39637-99-5 | C10H8ClF3O2 | 252.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 264122500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS39637-99-5
IUPAC Name(2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Molecular FormulaC10H8ClF3O2
InChI KeyPAORVUMOXXAMPL-VIFPVBQESA-N
SMILESCO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.0 %
Specific optical rotation-128° to -134° (20°C, 589 nm) (c=1, CHCl3)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only