trans-Zeatin (synthetic), 97+%
trans-Zeatin (synthetic), 97+%
trans-Zeatin (synthetic), 97+%
Thermo Scientific Chemicals

trans-Zeatin (synthetic), 97+%

CAS: 1637-39-4 | C10H13N5O | 219.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
100 mg
Packaging:
Glass bottle
Catalog number 264290100
Price (USD)
93.65
Online exclusive
104.00
Save 10.35
Each
Quantity:
10 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
93.65
Online exclusive
104.00
Save 10.35
Each
Chemical Identifiers
CAS1637-39-4
IUPAC Name(2E)-2-methyl-4-[(7H-purin-6-yl)amino]but-2-en-1-ol
Molecular FormulaC10H13N5O
InChI KeyUZKQTCBAMSWPJD-FARCUNLSSA-N
SMILESCC(CO)=C/CNC1=C2NC=NC2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Infrared spectrumConforms
Melting point>=207°C
Related substances=<2 % (HPLC)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only