Acetonitrile, 99.8%, for HPLC
Acetonitrile, 99.8%, for HPLC
Acetonitrile, 99.8%, for HPLC
Thermo Scientific Chemicals

Acetonitrile, 99.8%, for HPLC

CAS: 75-05-8 | C2H3N | 41.053 g/mol
Quantity:
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Catalog number 268270040
Price (JPY)
-
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.0002 %
UVat 240 nm A: =<0.015
UVat 260 nm A: =<0.005
UVat 280 nm A: =<0.005
Filtered to 0.2 micrometerFiltered
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only