Tetrahydrofuran, 99.8%, for HPLC, unstabilized
Tetrahydrofuran, 99.8%, for HPLC, unstabilized
Tetrahydrofuran, 99.8%, for HPLC, unstabilized
Thermo Scientific Chemicals

Tetrahydrofuran, 99.8%, for HPLC, unstabilized

Green features
CAS: 109-99-9, C4H8O, 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
4 L
Catalog number 268290010
Price (USD)
160.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
160.00
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.03 %
UVat 240 nm A: =<0.26
UVat 270 nm A: =<0.04
Peroxides=<0.0075 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only