2,2,4-Trimethylpentane, 99.5%, for HPLC
2,2,4-Trimethylpentane, 99.5%, for HPLC
2,2,4-Trimethylpentane, 99.5%, for HPLC
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99.5%, for HPLC

Green features
For HPLC | CAS: 540-84-1 | C8H18 | 114.232 g/mol
数量:
1 L
2.5 L
パッケージ:
Glass Bottle
製品番号(カタログ番号) 268810010
価格(JPY)
-
数量:
1 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS540-84-1
IUPAC Name2,2,4-trimethylpentane
Molecular FormulaC8H18
InChI KeyNHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILESCC(C)CC(C)(C)C
さらに表示
Appearance (Form)Liquid
Residue after evaporation=<0.0002 %
UVat 210 nm A: =<0.2
UVat 230 nm A: =<0.02
UV1 cm cell vs HPLC-grade water
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only