TheraPure™ 2'-MOE-T Phosphoramidite, 28-400 finish bottle
Thermo Scientific™

TheraPure™ 2'-MOE-T Phosphoramidite, 28-400 finish bottle

2'-MOE-T CEP is a phosphoramidite containing 5' DMT and the nucleoside thymidine (T, or 5-me-U) with 2'-O-methoxy-ethyl (MOE) protection, forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
27-1021-111 g
27-1021-822 g
Catalog number 27-1021-11
Price (TWD)
0.01
Each
Quantity:
1 g
Price (TWD)
0.01
Each

2'-MOE-T CEP is a phosphoramidite containing 5' DMT and the nucleoside thymidine (T, or 5-me-U) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methoxy-ethyl (MOE) modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 100-mL amber bottle with a 28-400 finish neck style, which is compatible with most MerMade synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-O-methoxyethyl-thymidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C43H55N5O10P
• Base—T
• Formula weight—819.0 g/mol
• CAS number—163878-63-5
• HPLC purity—≥99%; M-11 Impurity ≤0.01%
31P NMR purity—≥99%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)MerMade Synthesizers
FormPowder
GradeTheraPure™
Nucleic Acid TypeRNA
Nucleoside TypeThymidine (T)
Product Type2in-MOE-T CEP
Purity99%
Quantity1 g
Reagent Features2in-MOE modified
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Product LineTheraPure
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).