T Phosphoramidite, standard grade, 20-400 finish bottle
Thermo Scientific™

T Phosphoramidite, standard grade, 20-400 finish bottle

T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis. DNA phosphoramidites深入閱讀
Have Questions?
產品號碼Quantity
27-1736-112 g
產品號碼 27-1736-11
價格 (TWD)
940.00
Each
Quantity:
2 g
價格 (TWD)
940.00
Each

T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups (except T and U) designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30 mL Amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-deoxythymidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C40H49N4O8P
• Base—T
• Formula weight—744.82 g/mol
• CAS number—98796-51-1
• HPLC Purity—≥98.0%
31P NMR Purity—≥98.0%

For Research Use Only. Not for use in diagnostic procedures.
規格
For Use With (Equipment)Expedite Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeDNA
Nucleoside TypeDeoxythymidine (dT)
Product TypeT CEP
Purity98%
Quantity2 g
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
內容物與存放
Store in freezer (at or below -20°C).