2'-OMe-iBu-G Phosphoramidite, standard grade, 20-400 finish bottle
Thermo Scientific™

2'-OMe-iBu-G Phosphoramidite, standard grade, 20-400 finish bottle

2'-OMe-iBu-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methyl protection, for use in oligonucleotide詳細を見る
製品番号(カタログ番号)数量
27-1846-101 g
製品番号(カタログ番号) 27-1846-10
価格(JPY)
-
数量:
1 g

2'-OMe-iBu-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30-mL amber bottle with a 20-400 finish neck style, which is compatible with most Expedite synthesizers.

Features:
• HPLC purity—≥98.0%; Sum of non-primary peaks 140-152 ppm: ≤0.5%

研究用途にのみご使用ください。診断目的には使用できません。
仕様
31P NMR純度≥98.0%
CAS150780-67-9
化学物質名または材質5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methyl-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
概要Base: G
Base Protection: iBu
使用対象(アプリケーション)Oligonucleotide synthesis
使用対象 (装置)Expedite Synthesizers
形状Powder
フォーマットAmber bottle (20-400 finish neck style)
グレードStandard
分子式C45H56N7O9P
分子量869.9 g/mol
核酸タイプRNA
ヌクレオシドタイプGuanosine (G)
製品タイプ2in-OMe-iBu-G CEP
純度≥98.0% (HPLC)
数量1 g
試薬の特長2in-OMe modified
試薬タイプPhosphoramidite
出荷条件Room Temperature
Unit SizeEach
組成および保存条件
Store in freezer (at or below -20°C).