TheraPure™ Bz-dA Phosphoramidite, serum vial bottle
Thermo Scientific™

TheraPure™ Bz-dA Phosphoramidite, serum vial bottle

Bz-dA CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyadenosine (dA) with Benzoyl (Bz) base protection, for use詳細を見る
製品番号(カタログ番号)数量
27-2030-072 g
27-2030-121 g
製品番号(カタログ番号) 27-2030-07
価格(JPY)
-
数量:
2 g

Bz-dA CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyadenosine (dA) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The TheraPure trademark denotes a highest level of phosphoramidite purity, developed with the highest stringency of of high definiteion analytics and tightly controlled impurity profiles. Ideal for any therapeutic application. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N6-benzoyl-2'-deoxyadenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C47H52N7O7P
• Base—dA
• Base Protection—Bz
• Formula weight—857.95 g/mol
• CAS number—98796-53-3
• HPLC Purity—≥99%; M-11 Impurity ≤0.01%; Single critical impurity ≤0.3%; Presence of dC, dG, or T impurity ≤0.03%
31P NMR Purity—≥99%; Sum of non-primary peaks at 140-152 ppm: ≤0.3%; No peak(s) above 170 ppm; HMT impurity (∼146ppm): ≤0.10%

For Research Use Only. Not for use in diagnostic procedures.
仕様
使用対象 (装置)AKTA Synthesizers, Applied Biosystems Synthesizers
形状Powder
グレードTheraPure™
核酸タイプDNA
ヌクレオシドタイプDeoxyadenosine (dA)
製品タイプBz-dA CEP
純度99%
数量2 g
試薬タイプPhosphoramidite
出荷条件Room Temperature
製品ラインTheraPure
Unit SizeEach
組成および保存条件
Store in freezer (at or below -20°C).