Magnesium acetate tetrahydrate, 99.5%, for analysis
Magnesium acetate tetrahydrate, 99.5%, for analysis
Magnesium acetate tetrahydrate, 99.5%, for analysis
Magnesium acetate tetrahydrate, 99.5%, for analysis
Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, 99.5%, for analysis

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol
数量:
5 g
5 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 270500051
価格(JPY)
-
見積もりを依頼する
数量:
5 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16674-78-5
IUPAC Namemagnesium(2+) diacetate tetrahydrate
Molecular FormulaC4H14MgO8
InChI KeyXKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILESO.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
さらに表示
Titration Complexometric>=99.4 %
Total nitrogen compounds=<0.001 % (as N)
Barium (Ba)=<10 ppm
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<5 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only