2-Propylpentanoic acid, sodium salt, 98%
2-Propylpentanoic acid, sodium salt, 98%
2-Propylpentanoic acid, sodium salt, 98%
Thermo Scientific Chemicals

2-Propylpentanoic acid, sodium salt, 98%

CAS: 1069-66-5 | C8H15NaO2 | 166.2 g/mol
製品番号(カタログ番号) 271830050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1069-66-5
IUPAC Namesodium 2-propylpentanoate
Molecular FormulaC8H15NaO2
InChI KeyAEQFSUDEHCCHBT-UHFFFAOYSA-M
SMILES[Na+].CCCC(CCC)C([O-])=O
さらに表示
Appearance (Color)White
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only