Nalgene™ Dropper Bottles
Nalgene™ Dropper Bottles
Nalgene™ Dropper Bottles
Nalgene™ Dropper Bottles
Nalgene™ Dropper Bottles
Thermo Scientific™

Nalgene™ Dropper Bottles

Use for accurate, reliable, repeatable dispensing of various reagents. These dropper bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. An excellent alternative to pipetting devices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Color
2751-905015 mLWhite
2751-91254 mLWhite
2751-90258 mLWhite
2753-905015 mLAssorted
Catalog number 2751-9050
Price (USD)
60.58
Online Exclusive
71.25
Save 10.67 (15%)
Case of 25
Capacity (Metric):
15 mL
Closure Color:
White
Price (USD)
60.58
Online Exclusive
71.25
Save 10.67 (15%)
Case of 25

Choose Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles for accurate, reliable, repeatable dispensing of various reagents and as an excellent alternative to pipetting devices. Nalgene™ Dropper Bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. Delivers 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Excellent chemical resistance; suitable for most biotech, diagnostic and pharmaceutical applications.

  • Opaque white bottle ideal for UV lightsensistive solutions
  • Dropper control tip snaps into place for a secure fit
  • Dispensing tip and closures included; choose white or multi-colored closures
  • Bottles deliver 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time
  • Leakproof† when used with provided Nalgene tip and closure

Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Closure ColorWhite
Closure Size15-415 mm
Closure TypeDropper Control Tip
DescriptionLDPE Dropper Bottle
IncludesDispensing tip and closures
No. per Case25
No. per Pack25
Through the Closure DispensingYes
Capacity (English)0.5 oz.
Capacity (Metric)15 mL
Certifications/ComplianceISO 13485:2003
Closure IncludedYes
ColorWhite
Drop Size40 μL
Dropper Capacity15 mL
MaterialLDPE
Product TypeDropping Bottle
ShapeRound
Unit SizeCase of 25