1,5-Bis(methylamino)-3-oxapentane, 98%
1,5-Bis(methylamino)-3-oxapentane, 98%
1,5-Bis(methylamino)-3-oxapentane, 98%
Thermo Scientific Chemicals

1,5-Bis(methylamino)-3-oxapentane, 98%

CAS: 2620-27-1 | C6H16N2O | 132.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 275860050
Price (EUR)
467,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
467,00
Each
Chemical Identifiers
CAS2620-27-1
IUPAC Namemethyl({2-[2-(methylazaniumyl)ethoxy]ethyl})azanium
Molecular FormulaC6H18N2O
InChI KeyVXPJBVRYAHYMNY-UHFFFAOYSA-P
SMILESC[NH2+]CCOCC[NH2+]C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only