3-chloro-2-chloromethyl-1-propene, 96%
3-chloro-2-chloromethyl-1-propene, 96%
3-chloro-2-chloromethyl-1-propene, 96%
Thermo Scientific Chemicals

3-chloro-2-chloromethyl-1-propene, 96%

CAS: 1871-57-4 | C4H6Cl2 | 124.99 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 277070050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1871-57-4
IUPAC Name3-chloro-2-(chloromethyl)prop-1-ene
Molecular FormulaC4H6Cl2
InChI KeyXJFZOSUFGSANIF-UHFFFAOYSA-N
SMILESClCC(=C)CCl
さらに表示
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=95 %
Appearance (Form)Liquid
Refractive index1.4750 to 1.4790 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only