Glycerol, pure, 83.5-88.5 wt% aqueous solution
Glycerol, pure, 83.5-88.5 wt% aqueous solution
Glycerol, pure, 83.5-88.5 wt% aqueous solution
Thermo Scientific Chemicals

Glycerol, pure, 83.5-88.5 wt% aqueous solution

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
製品番号(カタログ番号)数量パッケージ
2773600505 LPlastic bottle
製品番号(カタログ番号) 277360050
価格(JPY)
-
数量:
5 L
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
さらに表示
Assay83.5 to 88.5 %
Appearance (Form)Viscous liquid
Refractive index1.4490 to 1.4550 (20°C, 589 nm)
Appearance (Color)Clear colorless to almost colorless
Water12 to 16 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only