4-Bromo-2-fluorobenzonitrile, 99+%
4-Bromo-2-fluorobenzonitrile, 99+%
4-Bromo-2-fluorobenzonitrile, 99+%
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzonitrile, 99+%

CAS: 105942-08-3 | C7H3BrFN | 200.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 290490250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS105942-08-3
IUPAC Name4-bromo-2-fluorobenzonitrile
Molecular FormulaC7H3BrFN
InChI KeyHGXWRDPQFZKOLZ-UHFFFAOYSA-N
SMILESFC1=CC(Br)=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)White to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder and/or chunks
Melting point69°C to 72°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only