(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%
(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%
(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%
Thermo Scientific Chemicals

(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%

CAS: 93621-94-4 | C8H17NO2 | 159.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 293460010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS93621-94-4
IUPAC Name(2S,5S)-2,5-bis(methoxymethyl)pyrrolidin-1-ium
Molecular FormulaC8H18NO2
InChI KeyHDXYTIYVVNJFLU-YUMQZZPRSA-O
SMILESCOC[C@@H]1CC[C@@H](COC)[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Infrared spectrumConforms
Refractive index1.4470 to 1.4500 (20°C, 589 nm)
GC>=95 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only