CaptureSelect™ GM-CSF Affinity Matrix
Thermo Scientific™

CaptureSelect™ GM-CSF Affinity Matrix

Thermo Scientific™ CaptureSelect™ GM-CSF Affinity Matrix is suitable for the purification of granulocyte-macrophage colony-stimulating factor (GM-CSF), a cytokine that functions as a white blood cell growth factor.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
294347201010 mL
29434720055 mL
294347205050 mL
Número de catálogo 2943472010
Precio (MXN)
-
Cantidad:
10 mL

CaptureSelect GM-CSF Affinity Matrix is suitable for the purification of granulocyte-macrophage colony-stimulating factor (GM-CSF), a cytokine that functions as a white blood cell growth factor. The affinity matrix enables purification of this cytokine with high purity and yield in a single step. The CaptureSelect affinity ligand is immobilized on a high-capacity agarose matrix.

For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Cantidad10 mL
Condiciones de envíoTemperatura ambiente
Capacidad de unión>2 mg/mL
FormularioSuspensión líquida
Tamaño de partículas65 μm
Línea de productosCaptureSelect
TipoAffinity Matrix
Unit SizeEach
Contenido y almacenamiento
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light