2-Chloropropionyl chloride, 95%
2-Chloropropionyl chloride, 95%
2-Chloropropionyl chloride, 95%
Thermo Scientific Chemicals

2-Chloropropionyl chloride, 95%

CAS: 7623-09-8 | C3H4Cl2O | 126.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 295165000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7623-09-8
IUPAC Name2-chloropropanoyl chloride
Molecular FormulaC3H4Cl2O
InChI KeyJEQDSBVHLKBEIZ-UHFFFAOYNA-N
SMILESCC(Cl)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4355 to 1.4420 (20°C, 589 nm)
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only