Acetic acid, 99+%, extra pure
Acetic acid, 99+%, extra pure
Acetic acid, 99+%, extra pure
Thermo Scientific Chemicals

Acetic acid, 99+%, extra pure

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
25 L
Packaging:
View all
Metal drum
Catalog number 295310250
Price (EUR)
438,00
Each
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (EUR)
438,00
Each
Chemical Identifiers
CAS64-19-7
IUPAC Nameacetic acid
Molecular FormulaC2H4O2
InChI KeyQTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILESCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3700 to 1.3730 (20°C, 589 nm)
Water=<0.04 %
Infrared spectrumConforms
Residue after evaporation=<0.01 %
Titration with NaOH>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only