Methanesulfonamide, 98%
Methanesulfonamide, 98%
Methanesulfonamide, 98%
Thermo Scientific Chemicals

Methanesulfonamide, 98%

CAS: 3144-09-0 | CH5NO2S | 95.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 296111000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3144-09-0
IUPAC Namemethanesulfonamide
Molecular FormulaCH5NO2S
InChI KeyHNQIVZYLYMDVSB-UHFFFAOYSA-N
SMILESCS(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Melting point88°C to 92°C
Appearance (Form)Powder or crystals or flakes
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only