2,2,6-Trimethyl-4H-1,3-dioxin-4-one, 94%
2,2,6-Trimethyl-4H-1,3-dioxin-4-one, 94%
2,2,6-Trimethyl-4H-1,3-dioxin-4-one, 94%
Thermo Scientific Chemicals

2,2,6-Trimethyl-4H-1,3-dioxin-4-one, 94%

CAS: 5394-63-8 | C7H10O3 | 142.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 296911000
Price (EUR)
66,80
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
66,80
Each
Chemical Identifiers
CAS5394-63-8
IUPAC Name2,2,6-trimethyl-2,4-dihydro-1,3-dioxin-4-one
Molecular FormulaC7H10O3
InChI KeyXFRBXZCBOYNMJP-UHFFFAOYSA-N
SMILESCC1=CC(=O)OC(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4580 to 1.4650 (20°C, 589 nm)
GC>=92 %
Acetone=<6 %
Appearance (Color)Clear yellow to dark brown
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only