Brefeldin A, natural
Brefeldin A, natural
Brefeldin A, natural
Thermo Scientific Chemicals

Brefeldin A, natural

CAS: 20350-15-6 | C16H24O4 | 280.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
Catalog number 297140050
Price (TWD)
-
Quantity:
5 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20350-15-6
IUPAC Name(1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one
Molecular FormulaC16H24O4
InChI KeyKQNZDYYTLMIZCT-PNFJWZTBSA-N
SMILESC[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Melting point201°C to 207°C
HPLC>=98.0 %
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only