Product Image
Thermo Scientific Chemicals

1-Hydroxybenzotriazole hydrate, 98

CAS: 123333-53-9 | C6;H5;N3;O·xH2;O | 135.12 g/mol
製品番号(カタログ番号) 300561000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS123333-53-9
IUPAC Name1H-1,2,3-benzotriazol-1-ol
Molecular FormulaC6H5N3O
InChI KeyASOKPJOREAFHNY-UHFFFAOYSA-N
SMILESON1N=NC2=CC=CC=C12
さらに表示
Appearance (Form)Crystalline powder
Titration with NaOH97.5 to 101.5 % (On dry substance)
Infrared spectrumConforms
HPLC>=97.5 %
Water11 to 26 % (K.F.)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only