alpha-Terpineol, 97+%
alpha-Terpineol, 97+%
alpha-Terpineol, 97+%
Thermo Scientific Chemicals

alpha-Terpineol, 97+%

CAS: 98-55-5 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Catalog number 301610025
Price (USD)
183.65
Online exclusive
204.00
Save 20.35
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
183.65
Online exclusive
204.00
Save 20.35
Each
Chemical Identifiers
CAS98-55-5
IUPAC Name2-[(1R)-4-methylcyclohex-3-en-1-yl]propan-2-ol
Molecular FormulaC10H18O
InChI KeyWUOACPNHFRMFPN-VIFPVBQESA-N
SMILESCC1=CC[C@@H](CC1)C(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Viscous liquid or low melting crystalline mass or
Refractive index1.4800 to 1.4850 (20°C, 589 nm)
Appearance (Color)Clear colorless to white
GC>=97.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only