Bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, 97%
Bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, 97%
Bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, 97%
Thermo Scientific Chemicals

Bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, 97%

CAS: 132705-51-2 | C12H24BrN3P·F6P | 466.19 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 302130250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS132705-51-2
IUPAC Namebromotris(pyrrolidin-1-yl)phosphanium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC12H24BrF6N3P2
InChI KeyCYKRMWNZYOIJCH-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.Br[P+](N1CCCC1)(N1CCCC1)N1CCCC1
さらに表示
Appearance (Form)Powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point>=100°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only