3,5-Bis(trifluoromethyl)benzylamine, 95%
3,5-Bis(trifluoromethyl)benzylamine, 95%
3,5-Bis(trifluoromethyl)benzylamine, 95%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzylamine, 95%

CAS: 85068-29-7 | C9H7F6N | 243.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 305640050
Price (EUR)
109,65
Online exclusive
122,00
Save 12,35 (10%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
109,65
Online exclusive
122,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS85068-29-7
IUPAC Name[3,5-bis(trifluoromethyl)phenyl]methanaminium
Molecular FormulaC9H8F6N
InChI KeyDHVHORCFFOSRBP-UHFFFAOYSA-O
SMILES[NH3+]CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or mass and/or
GC>=94 %
Appearance (Form)chunks
Infrared spectrumConforms
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only