1-Bromo-3-methyl-2-butene, 96%
1-Bromo-3-methyl-2-butene, 96%
1-Bromo-3-methyl-2-butene, 96%
Thermo Scientific Chemicals

1-Bromo-3-methyl-2-butene, 96%

CAS: 870-63-3 | C5H9Br | 149.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 307410500
Price (USD)
170.65
Online exclusive
189.00
Save 18.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
170.65
Online exclusive
189.00
Save 18.35
Each
Chemical Identifiers
CAS870-63-3
IUPAC Name1-bromo-3-methylbut-2-ene
Molecular FormulaC5H9Br
InChI KeyLOYZVRIHVZEDMW-UHFFFAOYSA-N
SMILESCC(C)=CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Argentometric95 to 105 % (ex Br)
Refractive index1.4880 to 1.4920 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only