3,5-Bis(trifluoromethyl)bromobenzene, 99%
3,5-Bis(trifluoromethyl)bromobenzene, 99%
3,5-Bis(trifluoromethyl)bromobenzene, 99%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)bromobenzene, 99%

CAS: 328-70-1 | C8H3BrF6 | 293.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 307700100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS328-70-1
IUPAC Name1-bromo-3,5-bis(trifluoromethyl)benzene
Molecular FormulaC8H3BrF6
InChI KeyCSVCVIHEBDJTCJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=CC(Br)=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4260 to 1.4280 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only