2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 95%
2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 95%
2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 95%
Thermo Scientific Chemicals

2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 95%

CAS: 29261-33-4 | C12F4N4 | 276.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3126700505 mgGlass bottle
Catalog number 312670050
Price (TWD)
-
Quantity:
5 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS29261-33-4
IUPAC Name2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
Molecular FormulaC12F4N4
InChI KeyIXHWGNYCZPISET-UHFFFAOYSA-N
SMILESFC1=C(F)C(=C(C#N)C#N)C(F)=C(F)C1=C(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow-green to orange-brown
Infrared spectrumConforms
Assay>=94 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only