Zinc trifluoromethanesulfonate, 98%
Zinc trifluoromethanesulfonate, 98%
Zinc trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Zinc trifluoromethanesulfonate, 98%

CAS: 54010-75-2 | C2F6O6S2Zn | 363.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 317090500
Price (USD)
179.65
Special offer
Online exclusive
Ends: 30-Jun-2026
211.00
Save 31.35 (15%)
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
179.65
Special offer
Online exclusive
Ends: 30-Jun-2026
211.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS54010-75-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to light grey
Infrared spectrumConforms
Titration Complexometric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only