Zinc acetate dihydrate, 98+%, ACS reagent
Zinc acetate dihydrate, 98+%, ACS reagent
Zinc acetate dihydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Zinc acetate dihydrate, 98+%, ACS reagent

CAS: 5970-45-6 | C4H6O4Zn·2H2O | 219.5 g/mol
数量:
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 317210025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5970-45-6
IUPAC Namezinc(2+) diacetate dihydrate
Molecular FormulaC4H10O6Zn
InChI KeyBEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILESO.O.[Zn++].CC([O-])=O.CC([O-])=O
さらに表示
Appearance (Form)Crystals
Titration Complexometric98.0 to 101.0 %
Insoluble matter=<0.005 %
Chloride (Cl)=<5 ppm
Magnesium (Mg)=<0.005 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only