Gallium, 99.99%, (trace metal basis)
Gallium, 99.99%, (trace metal basis)
Gallium, 99.99%, (trace metal basis)
Thermo Scientific Chemicals

Gallium, 99.99%, (trace metal basis)

CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Catalog number 317990500
Price (USD)
258.65
Special offer
Online exclusive
Ends: 28-Dec-2026
304.00
Save 45.35
Each
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
258.65
Special offer
Online exclusive
Ends: 28-Dec-2026
304.00
Save 45.35
Each
Chemical Identifiers
CAS7440-55-3
IUPAC Namegallium
Molecular FormulaGa
InChI KeyGYHNNYVSQQEPJS-UHFFFAOYSA-N
SMILES[Ga]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver white to silver grey
Total trace metal impurities=<0.01 %
Appearance (Form)Metallic solid or liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only