Osmium tetroxide, 4 wt.% solution in water
Osmium tetroxide, 4 wt.% solution in water
Osmium tetroxide, 4 wt.% solution in water
Thermo Scientific Chemicals

Osmium tetroxide, 4 wt.% solution in water

CAS: 20816-12-0 | O4Os | 254.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
10 mL
Packaging:
Glass bottle
Catalog number 319010100
Price (USD)
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20816-12-0
IUPAC Nametetraoxoosmium
Molecular FormulaO4Os
InChI KeyVUVGYHUDAICLFK-UHFFFAOYSA-N
SMILESO=[Os](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Strength3.92 to 4.08 %
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only