Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays
実際の製品は異なる場合があります
Thermo Scientific™

Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays

Easily handle these narrow-mouth HDPE bottles that are ready to feed into filling lines for various reagents, and conveniently packaged in shrink-wrap trays.
製品番号(カタログ番号)容量(メートル法)クロージャーサイズ
322089-0008250 mL24-415 mm
322089-000130 mL20-415 mm
322089-000260 mL20-415 mm
製品番号(カタログ番号) 322089-0008
価格(JPY)
71,640
キャンペーン価格
Ends: 26-Jun-2026
89,540
割引額 17,900 (20%)
Case of 300
容量(メートル法):
250 mL
クロージャーサイズ:
24-415 mm
Easily handle Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles, conveniently packaged in rigid, lint-free shrink-wrap trays and ready to feed into filling lines for various reagents. High density polyethylene bottles include polypropylene closures and rigid, lint-free trays.
Features:
  • Excellent impact and chemical resistance
  • Include opaque amber polypropylene closures
  • Translucent amber bottles provide light protection with the ability to see liquid level
  • Leakproof† when used with Nalgene closures

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

仕様
容量8 oz., 250 mL
キャップ色Natural
クロージャー材質Polypropylene
クロージャーサイズ24-415 mm
キャップタイプScrew (Bulk Packed, Unassembled)
概要Narrow-Mouth Packaging Bottles with Closures
直径(メートル法)61 mm
直径(メートル法)内部ネック18 mm
高さ(ヤードポンド法)4.09 in.
高さ(メートル法)104 mm
ネック仕上げ24 mm
箱あたりの入数300
パックあたり入数30
クロージャー分注によるNo
容量(ヤードポンド法)8 oz.
容量(メートル法)250 mL
認証/コンプライアンスManufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
Natural
材料HDPE
Narrow
製品タイプNarrow Mouth Bottle
形状Round
Unit SizeCase of 300