3-Bromopyruvic acid hydrate, 98%
3-Bromopyruvic acid hydrate, 98%
3-Bromopyruvic acid hydrate, 98%
Thermo Scientific Chemicals

3-Bromopyruvic acid hydrate, 98%

CAS: 206860-50-6 | C3H3BrO3 | 166.96 g/mol
Have Questions?
Catalog NumberQuantity
32569025025 g
Catalog number 325690250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS206860-50-6
IUPAC Name3-bromo-2-oxopropanoic acid
Molecular FormulaC3H3BrO3
InChI KeyPRRZDZJYSJLDBS-UHFFFAOYSA-N
SMILESOC(=O)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to light brown
Infrared spectrumConforms
Appearance (Form)Powder
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only