Methanol, 99.9%, for HPLC gradient grade
Methanol, 99.9%, for HPLC gradient grade
Methanol, 99.9%, for HPLC gradient grade
Thermo Scientific Chemicals

Methanol, 99.9%, for HPLC gradient grade

Green features
CAS: 67-56-1, CH4O, 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 325740025
Price (USD)
135.65
Online exclusive
150.00
Save 14.35
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
135.65
Online exclusive
150.00
Save 14.35
Each
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Filtered to 0.2 micrometerFiltered
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only