Dodecylbenzenesulfonic acid, sodium salt, technical
Dodecylbenzenesulfonic acid, sodium salt, technical
Dodecylbenzenesulfonic acid, sodium salt, technical
Thermo Scientific Chemicals

Dodecylbenzenesulfonic acid, sodium salt, technical

CAS: 25155-30-0 | C18H29NaO3S | 348.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 325910010
Price (EUR)
42,65
Online exclusive
47,50
Save 4,85 (10%)
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
42,65
Online exclusive
47,50
Save 4,85 (10%)
Each
Chemical Identifiers
CAS25155-30-0
IUPAC Namesodium 2-dodecylbenzene-1-sulfonate
Molecular FormulaC18H29NaO3S
InChI KeyHFQQZARZPUDIFP-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Appearance (Form)Powder or crystalline powder
AppearanceMay contain dark particles
Infrared spectrumConforms
Assay78 to 80 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only