Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
Thermo Scientific Chemicals

Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free

CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 326720025
Price (USD)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Boiling point40°C to 60°C
Acidity (CH3COOH)=<0.003 %
Residue after evaporation=<0.0002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only