Isopropanol, 99.8%, Extra Dry, AcroSeal™
Isopropanol, 99.8%, Extra Dry, AcroSeal™
Isopropanol, 99.8%, Extra Dry, AcroSeal™
Acros Organics

Isopropanol, 99.8%, Extra Dry, AcroSeal™

CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Catalog number 326960025
HSN 29051220
Price (INR)
39,271.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (INR)
39,271.00
Each
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=99.75 %
Water=<0.005 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only