Tris(hydroxymethyl)aminomethane, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Tris(hydroxymethyl)aminomethane, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Tris(hydroxymethyl)aminomethane, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, 99.85%, for molecular biology, DNAse, RNAse and Protease free

CAS: 77-86-1 | C4H11NO3 | 121.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 327360010
Price (USD)
145.65
Online exclusive
162.00
Save 16.35
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
145.65
Online exclusive
162.00
Save 16.35
Each
Specifications
Absorbance(0.1M in water), (0.1 M in water) at 0nm, 0.03 max. at 260nm
Assay Percent Range99.85%
Beilstein04, 303
CAS77-86-1
ChEBICHEBI:9754
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only