Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free

CAS: 57-13-6 | CH4N2O | 60.06 g/mol
数量:
1 kg
5 kg
パッケージ:
Glass Bottle
製品番号(カタログ番号) 327380050
価格(JPY)
-
数量:
5 kg
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
さらに表示
Heavy metals (as Pb)=<10 ppm
UVat 280 nm A: =<0.05
Chloride (Cl)=<5 ppm
Assay>=99.5 %
UV(8 M in water)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only