Bicine, 99+%, for biochemistry
Bicine, 99+%, for biochemistry
Bicine, 99+%, for biochemistry
Thermo Scientific Chemicals

Bicine, 99+%, for biochemistry

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 327711000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS150-25-4
IUPAC Namesodium 2-[bis(2-hydroxyethyl)amino]acetate
Molecular FormulaC6H12NNaO4
InChI KeyMFBDBXAVPLFMNJ-UHFFFAOYSA-M
SMILES[Na+].OCCN(CCO)CC([O-])=O
さらに表示
Appearance (Color)White
Infrared spectrumConforms
Water=<1 % (K.F.)
Titration with NaOH>=99.0 %
Heavy metals (as Pb)=<5 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only