Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
Thermo Scientific Chemicals

Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400

CAS: 25067-59-8 | (C14H11N)n | 193.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Catalog number 329690250
Price (USD)
476.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
476.00
Each
Chemical Identifiers
CAS25067-59-8
Molecular Formula(C14H11N)n
SMILES*-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2
MDL NumberMFCD00134336
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only