Diethyl (4-nitrobenzyl)phosphonate, 98%
Diethyl (4-nitrobenzyl)phosphonate, 98%
Diethyl (4-nitrobenzyl)phosphonate, 98%
Thermo Scientific Chemicals

Diethyl (4-nitrobenzyl)phosphonate, 98%

CAS: 2609-49-6 | C11H16NO5P | 273.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 332290050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2609-49-6
IUPAC Namediethyl [(4-nitrophenyl)methyl]phosphonate
Molecular FormulaC11H16NO5P
InChI KeyFORMFFDDQMCTCT-UHFFFAOYSA-N
SMILESCCOP(=O)(CC1=CC=C(C=C1)[N+]([O-])=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5220 to 1.5260 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear yellow to orange to dark red
NMR>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only