Thermo Scientific Chemicals

Ethyl 3-phenylglycidate, 90%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 332652500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 3-phenylglycidate, 90%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 332652500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 332652500
Price (USD)
Price: 170.00
Online price: 146.65
Your price:
Quantity
-

Chemical Identifiers

CAS
121-39-1
IUPAC Name
ethyl 3-phenyloxirane-2-carboxylate
Molecular Formula
C11H12O3
InChI Key
GOMAKLPNAAZVCJ-UHFFFAOYNA-N
SMILES
CCOC(=O)C1OC1C1=CC=CC=C1
Infrared spectrum
Conforms
Refractive index
1.5165 to 1.5205 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
GC
>=88 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text