Thermo Scientific Chemicals

Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals

Catalog number: 333110500
50 mL, Each
Thermo Scientific Chemicals

Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals

Catalog number: 333110500
50 mL, Each
Quantity
Catalog number: 333110500
Price (USD)
Quantity
-

Chemical Identifiers

CAS
87413-09-0, 75-09-2
Appearance (Color)
Colorless to light yellow
Appearance
May develop some turbidity or precipitate
Appearance (Form)
Solution
Titration Iodometric
10 to 15 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text