2-Methyl-1-buten-3-yne, 98%
2-Methyl-1-buten-3-yne, 98%
2-Methyl-1-buten-3-yne, 98%
Thermo Scientific Chemicals

2-Methyl-1-buten-3-yne, 98%

CAS: 78-80-8 | C5H6 | 66.1 g/mol
製品番号(カタログ番号) 336480100
価格(JPY)
-
数量:
10 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS78-80-8
IUPAC Name2-methylbut-1-en-3-yne
Molecular FormulaC5H6
InChI KeyBOFLDKIFLIFLJA-UHFFFAOYSA-N
SMILESCC(=C)C#C
さらに表示
Refractive index1.4145 to 1.4215 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only