Tetraacetylethylenediamine, 90% - FAQs
View additional product information for Tetraacetylethylenediamine, 90% - FAQs (344590500, 344592500)
1 product FAQs found
Is this Tetraacetylethylenediamine (TAED), 90% (Cat. Nos. 344592500, 344590500) uncoated ?
Yes, this Tetraacetylethylenediamine, 90% is uncoated.