1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, Thermo Scientific Chemicals
1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, Thermo Scientific Chemicals
1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, Thermo Scientific Chemicals

CAS: 95464-05-4 | Molecular Formula: C35H30Cl4FeP2Pd | Molecular Weight: 816.64 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 348680250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS95464-05-4
IUPAC NameBis(diphenylphosphino)ferrocene]palladium(II) chloride
Molecular FormulaC35H30Cl4FeP2Pd
SMILES[Fe].ClCCl.Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)816.64
さらに表示
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Carbon (C)50 to 53 %
Palladium (Pd)12.6 to 13.4 %
Appearance (Color)Orange to red-brown

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only